Learn More
Wnt-4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-20909
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Wnt-4 Polyclonal specifically detects Wnt-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
| Wnt-4 | |
| Polyclonal | |
| WNT4 | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Western Blot 1:500-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |