Learn More
Invitrogen™ WNT3A Monoclonal Antibody (1E6G4)
Mouse Monoclonal Antibody
Marke:  Invitrogen™ MA531954
Artikelnummer. 16325384
  • 506.00 € / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
MA5-31954 has been tested in indirect ELISA.
Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
WNT3A
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P56704
WNT3A
Purified recombinant fragment of human WNT3A (AA: 170-352) expressed in E. coli.
100 μL
Primary
Human
Antibody
IgG2b
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1E6G4
Unconjugated
WNT3A
MGC119418; MGC119419; MGC119420; Protein Wnt-3a; vestigial tail; vt; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3A; Wnt 3a; Wnt family member 3A; wnt[a]; wnt3; wnt3 l; wnt3 like; Wnt3a; Wnt-3a; WNT3A protein; wnt3l; Zwnt[a]
Mouse
Protein G
RUO
89780
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.