missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT6 Polyclonal Antibody

Beschreibung
WNT6 Polyclonal Antibody for Western Blot, IHC (F)
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is overexpressed in cervical cancer cell line and strongly coexpressed with another family member, WNT10A, in colorectal cancer cell line. The gene overexpression may play key roles in carcinogenesis. This gene and the WNT10A gene are clustered in the chromosome 2q35 region. The protein encoded by this gene is 97% identical to the mouse Wnt6 protein at the amino acid level.
Spezifikation
Spezifikation
| Antigen | WNT6 |
| Anwendungen | Immunohistochemistry (Frozen), Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.25 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 0.1% sodium azide; pH 7.4 |
| Gen | WNT6 |
| Gen-Zugriffsnummer | 0 |
| Gen-Alias | AA409270; Protein Wnt-6; wingless-related MMTV integration site 6; wingless-related MMTV integration site 6 homolog; wingless-type MMTV integration site family member 6; wingless-type MMTV integration site family, member 6; Wnt family member 6; WNT6; Wnt-6 |
| Gensymbole | WNT6 |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?