Learn More
Upchurch Scientific X10 Fitting flangeless for 1/16 inch O.D. tubingPEEK(TM) 1/4inch-28 flat bottom natural/blue
Marke:  Upchurch Scientific 02014-97
X10 Fitting flangeless for 1/16 inch O.D. tubingPEEK(TM) 1/4inch-28 flat bottom natural/blue
Artikelnummer. 12982119
  • 137.00 € / 10 Stück