Learn More
Medicom X100 Snooded cap PP 40 g/m² Extra White, in dispenser box
Marke:  Medicom 70.401
X100 Snooded cap PP 40 g/m² Extra White, in dispenser box
Artikelnummer. 16544121
  • ONLINE EXCLUSIVE PRICE
    23.43 €
    20.07 € / 100 Stück
    Sparen 3.36 €
    14% Rabatt
Verfügbar ab: 27-11-2026