Learn More
TERUMO Winged Infusion Safety Sets SURFLO™
Beschreibung
- Made out of kink resistant material to prevent kinking during placement for a higher patient comfort
- The wings are large and flexible for a good grip and secure fixation
- Reduced pressure on the blood vessel wall due to slightly angled wings
- Easy activation of the safety mechanism, fingers tips stay away from the cannula at all times
- Audible sound during activation of the safety mechanism
Spezifikation
Spezifikation
| Produkttyp | Infusion Set |
| Farbe | Black |
| Material | PVC Tubing, Siliconized Needle |
| Kanülen-Gauge | 22 G |
| Sterilität | Sterile |
| Latexfrei | Yes |
| Länge (metrisch) Schlauch | 30 cm |