Learn More
Invitrogen™ XLF Recombinant Rabbit Monoclonal Antibody (24GB480)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557512
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
XLF Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.
Spezifikation
| XLF | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NHEJ1 | |
| 1700029B21Rik; cernunnos; FLJ12610; Nhej1; non-homologous end joining factor 1; non-homologous end-joining factor 1; nonhomologous end-joining factor 1; protein cernunnos; Xlf; XRCC4-like factor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79840 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB480 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H9Q4 | |
| NHEJ1 | |
| A synthesized peptide derived from human XLF (aa 150-299). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |