missing translation for 'onlineSavingsMsg'
Learn More

XPG Antibody, Novus Biologicals™

Product Code. 18677714 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
100 μg
50 μg
Unit Size:
100 Mikroliter
50 Mikroliter
Product Code. Menge unitSize
18677714 50 μg 50 Mikroliter
18659661 100 μg 100 Mikroliter
2 options
This item is not returnable. View return policy

Product Code. 18677714

Brand: Novus Biologicals NBP29798450ul

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

XPG Polyclonal antibody specifically detects XPG in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen XPG
Anwendungen Immunofluorescence
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunocytochemistry/ Immunofluorescence 1:100-1:500
Zusammensetzung PBS (pH 7.0)
Gen-Alias COFS3, DNA excision repair protein ERCC-5, DNA repair protein complementing XP-G cells, EC 3.1, ERCM2, excision repair cross-complementing rodent repair deficiency, complementationgroup 5, excision repair protein, xeroderma pigmentosum complementation group G protein, Xeroderma pigmentosum group G-complementing protein, xeroderma pigmentosum, complementation group G, XPGC, XPG-complementing protein, XPGUVDR
Wirtsspezies Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human XPG fragment.
Reinigungsverfahren Antigen and protein A Affinity-purified
Menge 50 μg
Regulatorischer Status RUO
Forschungsgebiet Cancer, DNA Repair, Nucleotide Excision Repair
Primär oder sekundär Primary
Gen-ID (Entrez) 2073
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.