missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Yeast and Mold Agar
Beschreibung
Support the growth of yeasts and molds with Yeast and Mold Agar (YM), ideal for accurate fungal isolation and enumeration.
Spezifikation
Spezifikation
| Farbe | Transparent |
| Inhalt und Lagerung | 2°C to 12°C |
| Format | 90 mm |
| Herstellungsort | Germany |
| Produkttyp | Agar Plate |
| Füllvolumen | 17 mL |
| Menge | 10 Platten |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?