Learn More
YMC Group YMC-Pack ODS-A, Classical Analytical HPLC Column (4.6 mm ID), 12 nm, S-5 µm, 250 x 4.6 mm
Marke: YMC Group AA12S05-2546WT
YMC-Pack ODS-A, Classical Analytical HPLC Column (4.6 mm ID), 12 nm, S-5 µm, 250 x 4.6 mm