missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB17 Monoclonal Antibody (OTI7G8), TrueMAB™, OriGene
Description
ZBTB17 Monoclonal antibody specifically detects ZBTB17 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ZBTB17 |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI7G8 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gen | ZBTB17 |
| Gen-Zugriffsnummer | Q13105 |
| Gen-Alias | AA589413; LP-1; Miz1; Miz-1; Myc-interacting zinc finger protein 1; Myc-interacting Zn finger protein-1; mZ13; pHZ-67; Polyomavirus late initiator promoter-binding protein; ZBTB17; Zfp100; Zfp-100; zinc finger and BTB domain containing 17; zinc finger and BTB domain-containing protein 17; Zinc finger protein 100; zinc finger protein 151; zinc finger protein 151 (pHZ-67); zinc finger protein 60; Zinc finger protein Z13; ZNF151; ZNF60 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?