missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
ZEB2 Monoclonal antibody specifically detects ZEB2 in Human samples. It is validated for Immunohistochemistry,Flow Cytometry (Intracellular Staining),CyTOF
Spezifikation
Spezifikation
| Antigen | ZEB2 |
| Anwendungen | Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF |
| Klassifikation | Monoclonal |
| Klon | 923328 |
| Konjugat | Janelia Fluor 635 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | KIAA0569FLJ42816, SIP1HSPC082, Smad-interacting protein 1, SMADIP1ZFHX1BSIP-1, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b |
| Wirtsspezies | Mouse |
| Immunogen | E. coli-derived recombinant human ZEB2/SIP1, Asn363-Lys537, Accession # O60315 |
| Menge | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?