missing translation for 'onlineSavingsMsg'
Learn More

Zebularin, TRC

Product Code. 30451955
Change view
Click to view available options
Menge:
100 mg
Unit Size:
100 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30451955 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451955 Supplier Toronto Research Chemicals Supplier No. TRCZ276000100MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Zebularine
CAS 3690-10-6
Summenformel C9 H12 N2 O5
Menge 100 mg
Synonym 1-β-D-Ribofuranosyl-2(1H)-pyrimidinone, 4-Deoxyuridine, NSC 309132, Pyrimidin-2-one β-D-Ribofuranoside
InChI-Formel InChI=1S/C9H12N2O5/c12-4-5-6(13)7(14)8(16-5)11-3-1-2-10-9(11)15/h1-3,5-8,12-14H,4H2/t5-,6-,7-,8-/m1/s1
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC=NC2=O
IUPAC-Name 1-[(2R,3R,4S,5R)-3,4-DiHydroxy-5-(Hydroxymethyl)oxolan-2-yl]pyrimidin-2-on
Molekulargewicht (g/mol) 228.2
Formelmasse 228.0746
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.