Learn More
Invitrogen™ Zelminemab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA559683
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30282528
  • 500.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Zelminemab is a biosimilar that targets ADCYAP1R1.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Zelminemab Humanized
Recombinant Monoclonal
Unconjugated
AMG-301
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized