Beschreibung
- NH2P perfect for ion exchange applications
- Irregular Amorphous Silica Gel
- High Purity
- European Manufacture
- Swiss accuracy and consistency
- Excellent batch to batch reproducibility
- ISO Certification
- High Chemical and Mechanical Stability
- Narrow Particles Size Distribution
- 20 to 500 μm particles size range
- Available in 60 A or 90 A pore size
- High loadability
- Selection of pack sizes available
- Suitable for gravity, flash, prep and process scale
Spezifikation
Spezifikation
| Partikelgröße | 40 bis 63 μm |
| Porengröße | 60 Å |
| Verfahren | Chromatography |
| Menge | 1 kg |
| Typ | Silica Gel |
| Material | Silica Gel |
| Phase | NH2Pfunktionalisiert |
| Partikelform | Irregular |