missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA565397
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: ENYSNLLSVEV WKADDQMERD HRNPDEQARQ FLILKNQTPI EERGDLFGKA LNLNTDFVSL RQVPYK Highest antigen sequence identity to the following orthologs - mouse 70%, rat 76%.
Zinc finger proteins (Zfp) are encoded by a large family of genes present in many organisms including yeast and human. Some of them are transcriptional activators and bind specifically to DNA by zinc mediated folded structures commonly known as zinc fingers. The putative Zfp-1 (Zinc finger protein 1 homolog) protein contains in addition to 7 zinc fingers, two helix-turn-helix motifs.
Spezifikation
| ZFP1 | |
| Polyclonal | |
| Unconjugated | |
| ZFP1 | |
| FLJ34243; Fnp-1; mkr-1; Protein mKR1; RGD1565049; ZFP1; Zfp-1; ZFP1 zinc finger protein; zinc finger protein 1; Zinc finger protein 1 homolog; zinc finger protein 475; ZNF475 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 162239 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P2D0 | |
| ZFP1 | |
| Recombinant Human ZFP1. Recombinant protein control fragment (Product #RP-104802). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur