missing translation for 'onlineSavingsMsg'
Learn More

Zinkacetat-Dihydrat, TRC

Artikelnummer. 30452337
Ansicht ändern
Click to view available options
Menge:
1 g
5 g
10 g
Packungsgröße:
1 Gramm
10 Gramm
5 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
30452337 10 g 10 Gramm
30454703 5 g 5 Gramm
30456171 1 g 1 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452337 Supplier Toronto Research Chemicals Supplier No. TRCZ70156310G
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5970-45-6
Summenformel 2 C2 H3 O2 . Zn . 2 H2 O
Molekulargewicht (g/mol) 219.53
Synonym Acetic acid, zinc salt, hydrate (2:1:2), Acetic acid, zinc salt, dihydrate (8CI,9CI), Wilzin, Zinc acetate dihydrate, Zinc diacetate dihydrate
IUPAC-Name Zink; Diacetat; Dihydrat
SMILES O.O.[Zn+2].CC(=O)[O-].CC(=O)[O-]

Specifications

Chemischer Name oder Material Zinc Acetate Dihydrate
Menge 10 g
InChI-Formel InChI=1S/2C2H4O2.2H2O.Zn/c2*1-2(3)4;;;/h2*1H3,(H,3,4);2*1H2;/q;;;;+2/p-2
Formelmasse 217.9769
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.