Learn More
Invitrogen™ Zolbetuximab Chimeric Recombinant Human Monoclonal Antibody
Beschreibung
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Zolbetuximab is under investigation in clinical trial NCT01630083 (Efficacy and Safety of IMAB362 in Combination With the EOX Regimen for CLDN18. 2-positive Gastric Cancer).
Spezifikation
Spezifikation
| Antigen | Zolbetuximab Chimeric |
| Anwendungen | ELISA, Flow Cytometry |
| Klassifikation | Recombinant Monoclonal |
| Konzentration | 3.3 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with no preservative; pH 7.4 |
| Gen-Alias | claudiximab; GC-182; IMAB362; IMAB-362 |
| Wirtsspezies | Human |
| Immunogen | Human CLDN18. |
| Reinigungsverfahren | Protein A/G |
| Mehr anzeigen |