Learn More
Zyxin Antibody (CL2502) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-36768JF549
Artikelnummer. 30300440
  • 737.00 € / 0.10 Milliliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Zyxin Monoclonal antibody specifically detects Zyxin in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Zyxin
Monoclonal
Janelia Fluor 549
CTCL tumor antigen se14-3, CTCL-associated antigen se14-3, cutaneous T-cell lymphoma associated antigen se14-3, Cutaneous T-cell lymphoma-associated antigen se14-3, KIAA1125, MGC31836, predicted protein of HQ2893, PRKCBP1, PRO2893, protein kinase C-binding protein 1, Rack7, RACK7protein kinase C binding protein 1, zinc finger MYND domain containing protein 8, Zinc finger MYND domain-containing protein 8, zinc finger, MYND-type containing 8
This antibody was developed using a recombinant protein derived from Q15942, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
7791
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL2502
50mM Sodium Borate
Mouse
Protein A purified