CAS RN 156791-82-1
CAS RN 156791-82-1
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Hosenkoside A, MedChemExpress
MedChemExpress Hosenkoside A is a baccharane glycoside isolated from the seeds of impatiens balsamina.