CAS RN 2070009-49-1
CAS RN 2070009-49-1
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
BET-BAY 002 (S enantiomer), MedChemExpress
MedChemExpress BET-BAY 002 S enantiomer is the S-enantiomer of BET-BAY 002. BET-BAY 002 is a BET inhibitor.