CAS RN 239087-04-8
IUPAC Name:
[4-fluoro-3-(trifluoromethyl)phenyl](phenyl)methanone
Synonyme:
[4-fluoro-3-(trifluoromethyl)phenyl](phenyl)methanone
Molekulargewicht (g/mol):
268.21
Summenformel:
C14H8F4O
InChi Key:
BWBZTYREKMHBFH-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1C(F)(F)F)C(=O)C1=CC=CC=C1
1
–
1
von
1
Ergebnisse
1
4-Fluor-3-(trifluormethyl)benzophenon, 97+ %, Thermo Scientific™
CAS: 239087-04-8 Summenformel: C14H8F4O Molekulargewicht (g/mol): 268.211 MDL-Nummer: MFCD00061258 InChI-Schlüssel: BWBZTYREKMHBFH-UHFFFAOYSA-N Synonym: 4-fluoro-3-trifluoromethyl benzophenone,4-fluoro-3-trifluoromethyl phenyl phenyl methanone,acmc-1cehg,4-fluoro-3-trifluoromethyl phenyl phenyl ketone,4-fluoro-3-trifluoromethyl phenyl-phenylmethanone,methanone, 4-fluoro-3-trifluoromethyl phenyl phenyl PubChem CID: 2737556 IUPAC-Name: [4-fluor-3-(trifluormethyl)phenyl]-phenylmethanon SMILES: C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)F)C(F)(F)F