CAS RN 5680-86-4
CAS RN 5680-86-4
IUPAC Name:
5-(benzyloxy)-2-{[(benzyloxy)carbonyl]amino}-5-oxopentanoic acid
Synonyme:
5-(benzyloxy)-2-{[(benzyloxy)carbonyl]amino}-5-oxopentanoic acid
Molekulargewicht (g/mol):
371.39
Summenformel:
C20H21NO6
InChi Key:
TWIVXHQQTRSWGO-UHFFFAOYNA-N
SMILES:
OC(=O)C(CCC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
1
–
1
von
1
Ergebnisse
1
N-benzyloxycarbonyl-D-Glutaminsäure5-Benzylester, 95 %, Thermo Scientific™
CAS: 5680-86-4 Summenformel: C20H21NO6 Molekulargewicht (g/mol): 371.389 MDL-Nummer: MFCD00065697 InChI-Schlüssel: TWIVXHQQTRSWGO-KRWDZBQOSA-N Synonym: z-glu obzl-oh,cbz-glu obzl-oh,2s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid,5-benzyl n-carbobenzoxy-l-glutamate,n-cbz-l-glutamic acid 5-benzyl ester,s-2-benzyloxycarbonylamino-pentanedioic acid 5-benzyl ester,s-5-benzyloxy-2-benzyloxy carbonyl amino-5-oxopentanoic acid,z-glu obzl oh,pubchem14964,z-glu-obzl-oh PubChem CID: 3049860 IUPAC-Name: (2S)-5-Oxo-5-phenylmethoxy-2-(phenylmethoxycarbonylamino)pentansäure SMILES: C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2=CC=CC=C2