CAS RN 5947-49-9
CAS RN 5947-49-9
IUPAC Name:
(1S,4aS,10aR)-6-hydroxy-1,4a-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-1-carboxylic acid
Synonyme:
podocarpic acid
Molekulargewicht (g/mol):
274.36
Summenformel:
C17H22O3
InChi Key:
VJILEYKNALCDDV-OIISXLGYSA-N
SMILES:
C[C@@]1(CCC[C@@]2(C)[C@H]1CCC1=CC=C(O)C=C21)C(O)=O
1
–
1
von
1
Ergebnisse
1
Podocarpic acid, MedChemExpress
MedChemExpress Podocarpic acid is a natural product, which has the best all-round positive effect and acts as a novel TRPA1 activator.