CAS RN 100061-59-4
CAS RN 100061-59-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
2,6-Diamino-4-(benzyloxy)pyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,6-Diamino-4-(benzyloxy)pyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.