CAS RN 100434-10-4
CAS RN 100434-10-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
[3,4-Bis(benzyloxy)phenyl]-1,2-ethanediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[3,4-Bis(benzyloxy)phenyl]-1,2-ethanediol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.