CAS RN 10361-43-0
IUPAC Name:
bismuthanetriol
Synonyme:
bismuthanetriol
Molekulargewicht (g/mol):
263.03
Summenformel:
BiH6O3
InChi Key:
IAQAJTTVJUUIQJ-UHFFFAOYSA-N
SMILES:
O.O.O.[Bi]
1
–
2
von
2
Ergebnisse
1
Bismuthydroxid, tech., Thermo Scientific Chemicals
CAS: 10361-43-0 Summenformel: BiH3O3 Molekulargewicht (g/mol): 260.00 MDL-Nummer: MFCD00046188 InChI-Schlüssel: TZSXPYWRDWEXHG-UHFFFAOYSA-K IUPAC-Name: bismuth(3+) trihydroxide SMILES: [OH-].[OH-].[OH-].[Bi+3]
2
Bismuthydroxid, Thermo Scientific Chemicals
CAS: 10361-43-0 Summenformel: BiH3O3 Molekulargewicht (g/mol): 260.00 MDL-Nummer: MFCD00046188 InChI-Schlüssel: TZSXPYWRDWEXHG-UHFFFAOYSA-K IUPAC-Name: bismuth(3+) trihydroxide SMILES: [OH-].[OH-].[OH-].[Bi+3]