CAS RN 107-20-0
CAS RN 107-20-0
IUPAC Name:
2-chloroacetaldehyde
Synonyme:
chloroacetaldehyde rcra waste number P023 chloroacetaldehyde (substance) ccris 856 chloroaldehyde chloroacetaldehyde monomer
Molekulargewicht (g/mol):
78.5
Summenformel:
C2H3ClO
InChi Key:
QSKPIOLLBIHNAC-UHFFFAOYSA-N
SMILES:
ClCC=O