CAS RN 1070-34-4
CAS RN 1070-34-4
IUPAC Name:
4-ethoxy-4-oxobutanoic acid
Synonyme:
butanedioic acid, monoethyl ester
Molekulargewicht (g/mol):
146.14
Summenformel:
C6H10O4
InChi Key:
LOLKAJARZKDJTD-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCC(O)=O
1
–
2
von
2
Ergebnisse
1
Ethylwasserstoff-Succinat, 95 %, Thermo Scientific Chemicals
CAS: 1070-34-4 Summenformel: C6H10O4 Molekulargewicht (g/mol): 146.142 MDL-Nummer: MFCD00053346 InChI-Schlüssel: LOLKAJARZKDJTD-UHFFFAOYSA-N Synonym: monoethyl succinate,ethyl hydrogen succinate,mono-ethyl succinate,butanedioic acid, monoethyl ester,butanedioic acid, 1-ethyl ester,succinic acid monoethyl ester,monoethyl butanedioate,bernsteinsaureathylester,succinic acid monoethyl,succinic acid ethyl ester PubChem CID: 70610 IUPAC-Name: 4-Ethoxy-4-Oxobutansäure SMILES: CCOC(=O)CCC(=O)O