CAS RN 115144-35-9
CAS RN 115144-35-9
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
MedChemExpress D-Luciferin potassium
MedChemExpress D-Luciferin (D-(-)-Luciferin) potassium is the substrate of luciferases that catalyze the production of light in bioluminescent insects.