CAS RN 129-94-2
IUPAC Name:
2-[(8-chloronaphthalen-1-yl)sulfanyl]acetate
Synonyme:
2-[(8-chloronaphthalen-1-yl)sulfanyl]acetate
Molekulargewicht (g/mol):
251.7
Summenformel:
C12H8ClO2S
InChi Key:
WPNAGQJVWAFQJV-UHFFFAOYSA-M
SMILES:
[O-]C(=O)CSC1=C2C(Cl)=CC=CC2=CC=C1
1
–
1
von
1
Ergebnisse
1
2-(8-Chlor-1-naphthylthio)-Essigsäure, 98 %
CAS: 129-94-2 Summenformel: C12H8ClO2S Molekulargewicht (g/mol): 251.70 MDL-Nummer: MFCD00043901 InChI-Schlüssel: WPNAGQJVWAFQJV-UHFFFAOYSA-M Synonym: 8-chloro-1-naphthylthio acetic acid,2-8-chloro-1-naphthylthio acetic acid,2-8-chloronaphthalen-1-yl sulfanyl acetic acid,2-8-chloronaphthalen-1-yl thio acetic acid,8-chloronaphthalen-1-yl sulfanyl acetic acid,2-8-chloro-1-naphthyl thio acetic acid,maybridge1_001748,8-chloro-1-naphthyl sulfanyl acetic acid,acmc-1bugo,2-8-chloronaphthylthio acetic acid PubChem CID: 67219 IUPAC-Name: 2-(8-chlornaphthalen-1-yl)sulfanylEssigsäure SMILES: [O-]C(=O)CSC1=C2C(Cl)=CC=CC2=CC=C1