CAS RN 1309357-15-0
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
R&D Systems™ CX-4945
A Tocris product, part of R&D Systems. Potent and selective CK2α and CK2α' inhibitor. Also blocks CK2-dependent HIF-1α transcription, causes cell-cycle arrest, induces apoptosis in cancer cells and induces stem cell reprogramming
2
Silmitasertib sodium salt, MedChemExpress
MedChemExpress Silmitasertib sodium salt is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.