CAS RN 1343364-54-4
IUPAC Name:
trisodium [5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[(hydrogen phosphonatooxy)(oxido)phosphoryl]methyl}phosphonate
Synonyme:
trisodium [5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl [hydrogen phosphonatooxy(oxido)phosphoryl]methylphosphonate
Molekulargewicht (g/mol):
571.16
Summenformel:
C11H15N5Na3O12P3
InChi Key:
CBAGKCWFSRTVER-UHFFFAOYNA-K
SMILES:
[Na+].[Na+].[Na+].NC1=C2N=CN(C3OC(COP([O-])(=O)CP([O-])(=O)OP(O)([O-])=O)C(O)C3O)C2=NC=N1