CAS RN 13446-24-7
IUPAC Name:
dimagnesium(2+) (phosphonatooxy)phosphonate
Synonyme:
dimagnesium(2+) diphosphate
Molekulargewicht (g/mol):
222.55
Summenformel:
Mg2O7P2
InChi Key:
XZTWHWHGBBCSMX-UHFFFAOYSA-J
SMILES:
[Mg++].[Mg++].[O-]P([O-])(=O)OP([O-])([O-])=O
1
–
1
von
1
Ergebnisse
1
Magnesiumpyrophosphat, Thermo Scientific Chemicals
CAS: 13446-24-7 Summenformel: Mg2O7P2 Molekulargewicht (g/mol): 222.551 MDL-Nummer: MFCD00049482 InChI-Schlüssel: XZTWHWHGBBCSMX-UHFFFAOYSA-J Synonym: magnesium pyrophosphate,unii-al34y660jv,diphosphoric acid, magnesium salt 1:2,dimagnesium diphosphate,acmc-1but0,2mg.p2o7,magnesium pyrophosphate 50g,dimagnesium phosphonato phosphate,dimagnesium 2+ diphosphate,dimagnesium 2+ ion diphosphate PubChem CID: 122213 IUPAC-Name: Dimagnesium;phosphonatphosphat SMILES: [O-]P(=O)([O-])OP(=O)([O-])[O-].[Mg+2].[Mg+2]