CAS RN 14214-32-5
IUPAC Name:
1-[4-(4-methoxyphenoxy)phenyl]-3,3-dimethylurea
Synonyme:
difenoxuron
Molekulargewicht (g/mol):
286.33
Summenformel:
C16H18N2O3
InChi Key:
AMVYOVYGIJXTQB-UHFFFAOYSA-N
SMILES:
COC1=CC=C(OC2=CC=C(NC(=O)N(C)C)C=C2)C=C1