CAS RN 144-41-2
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Morphothion, TRC
CAS: 144-41-2 Summenformel: C8 H16 N O4 P S2 Molekulargewicht (g/mol): 285.32 Synonym: Phosphorodithioic acid, O,O-dimethyl S-(morpholinocarbonylmethyl) ester (6CI),Phosphorodithioic acid, O,O-dimethyl ester, S-ester with 4-(mercaptoacetyl)morpholine (7CI,8CI),Morpholine, 4-(mercaptoacetyl)-, S-ester with O,O-dimethyl phosphorodithioate (8CI),4-(Mercaptoacetyl)morpholine O,O-dimethyl phosphorodithioate,Ekatin F,Ekatin M,Morphothion,Morphotox,O,O-Dimethyl S-(morpholinocarbonylmethyl) phosphorodithioate IUPAC-Name: 2-dimethoxyphosphinothioylsulfanyl-1-morpholin-4-ylethanone SMILES: COP(=S)(OC)SCC(=O)N1CCOCC1