CAS RN 1460-34-0
IUPAC Name:
sodium 3-methyl-2-oxopentanoate
Synonyme:
sodium 3-methyl-2-oxopentanoate
Molekulargewicht (g/mol):
152.13
Summenformel:
C6H9NaO3
InChi Key:
SMDJDLCNOXJGKC-UHFFFAOYNA-M
SMILES:
[Na+].CCC(C)C(=O)C([O-])=O
1
–
1
von
1
Ergebnisse
1
MedChemExpress 3-Methyl-2-oxovaleric acid
MedChemExpress 3-Methyl-2-oxovaleric acid is a neurotoxin, an acidogen, and a metabotoxin, and also an abnormal metabolite that arises from the incomplete breakdown of branched-chain amino acids.