CAS RN 1494675-86-3
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
TBA-7371, MedChemExpress
MedChemExpress TBA-7371 is a potent, noncovalent DprE1 inhibitor. TBA-7371 has potent antitubercular activity .