CAS RN 159783-22-9
CAS RN 159783-22-9
IUPAC Name:
3,5-difluoropyridin-4-amine
Synonyme:
3,5-difluoropyridin-4-amine
Molekulargewicht (g/mol):
130.1
Summenformel:
C5H4F2N2
InChi Key:
XWAOYJHUAQFFMT-UHFFFAOYSA-N
SMILES:
NC1=C(F)C=NC=C1F
1
–
1
von
1
Ergebnisse
1
4-Amino-3,5-difluorpyridin, 97 %, Thermo Scientific Chemicals
CAS: 159783-22-9 Summenformel: C5H4F2N2 Molekulargewicht (g/mol): 130.1 InChI-Schlüssel: XWAOYJHUAQFFMT-UHFFFAOYSA-N PubChem CID: 2737654 IUPAC-Name: 3,5-Difluorpyridin-4-Amin SMILES: C1=C(C(=C(C=N1)F)N)F