CAS RN 1624117-53-8
CAS RN 1624117-53-8
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
XY1, MedChemExpress
MedChemExpress XY1 is a very close analogue of SGC707 (a potent, selective, and non-competitive inhibitor of PRMT3 with IC50 of 31 nM), but XY1 is completely inactive.