CAS RN 162929-64-8
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
3
von
3
Ergebnisse
1
MedChemExpress C-telopeptide
MedChemExpress C-telopeptide, a cross-linked peptide of type I collagen, is released during bone resorption and has been correlated with bone mineral density (BMD).
2
MedChemExpress C-telopeptide
MedChemExpress C-telopeptide, a cross-linked peptide of type I collagen, is released during bone resorption and has been correlated with bone mineral density (BMD).
3
MedChemExpress C-telopeptide
MedChemExpress C-telopeptide, a cross-linked peptide of type I collagen, is released during bone resorption and has been correlated with bone mineral density (BMD).