CAS RN 171783-05-4
CAS RN 171783-05-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
MedChemExpress Crosstide
MedChemExpress Crosstide is a peptide analog of glycogen synthase kinase α/β fusion protein sequence which is a substrate for Akt.
2
MedChemExpress Crosstide
MedChemExpress Crosstide is a peptide analog of glycogen synthase kinase α/β fusion protein sequence which is a substrate for Akt.