CAS RN 19220-93-0
IUPAC Name:
2,3,4,5,6-pentafluorophenyl acetate
Synonyme:
2,3,4,5,6-pentafluorophenyl acetate
Molekulargewicht (g/mol):
226.1
Summenformel:
C8H3F5O2
InChi Key:
ZXTVBLZVILLKPM-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F