CAS RN 194785-18-7
CAS RN 194785-18-7
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
MedChemExpress 6-ROX
MedChemExpress 6-ROX is a fluorescent oligonucleotide marker, acts as an acceptor molecule coupled to 5-FAM as the donor in FRET imaging.