CAS RN 202592-23-2
CAS RN 202592-23-2
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
JQ-1 (carboxylic acid), MedChemExpress
MedChemExpress JQ-1 carboxylic acid, a type of (+)-JQ-1 derivative, is a potent BET bromodomain inhibitor for downregulating PD-L1 expression on the surface of tumor cells. JQ-1 carboxylic acid can be used as a precursor to synthesize PROTACs, which targets BET bromodomains.