CAS RN 245445-88-9
CAS RN 245445-88-9
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
3
von
3
Ergebnisse
1
MedChemExpress T7 Tag Peptide
MedChemExpress T7 Tag Peptide is a protein tag derived from the N-terminal 11 residues of the major T7 capsid protein, gp 10. T7 Tag Peptide can be used in different immunoassays as well as affinity purification.
2
MedChemExpress T7 Tag Peptide
MedChemExpress T7 Tag Peptide is a protein tag derived from the N-terminal 11 residues of the major T7 capsid protein, gp 10. T7 Tag Peptide can be used in different immunoassays as well as affinity purification.
3
MedChemExpress T7 Tag Peptide
MedChemExpress T7 Tag Peptide is a protein tag derived from the N-terminal 11 residues of the major T7 capsid protein, gp 10. T7 Tag Peptide can be used in different immunoassays as well as affinity purification.