CAS RN 2457-80-9
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
5'-Deoxy-5'-(methylthio)adenosine, TRC
CAS: 2457-80-9 Summenformel: C11 H15 N5 O3 S Molekulargewicht (g/mol): 297.33 Synonym: 5'-S-Methyl-5'-thioadenosine,5'-(Methylthio)-5'-deoxyadenosine,5'-(Methylthio)adenosine,5'-Deoxy(methylthio)adenosine,5'-Deoxy-5'-(methylthio)adenosine,5'-S-Methylthioadenosine,NSC 335422,Vitamin L2,β-D-Ribofuranose, 1-(6-amino-9H-purin-9-yl)-1-deoxy-5-S-methyl-5-thio- IUPAC-Name: (2R,3R,4S,5S)-2-(6-aminopurin-9-yl)-5-(methylsulfanylmethyl)oxolane-3,4-diol SMILES: CSC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23
2
5'-Methylthioadenosine, MedChemExpress
MedChemExpress 5'-Methylthioadenosine (5'-(Methylthio)-5'-deoxyadenosine) is a nucleoside generated from S-adenosylmethionine (SAM) during polyamine synthesis.