CAS RN 245747-08-4
CAS RN 245747-08-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Pyr6, MedChemExpress
MedChemExpress Pyr6 is a selective inhibitor of TRPC3 with IC50 of 0.49 uM(Ca2+ influx inhibition in thapsigargin depleted native RBL-2H3 cells).