CAS RN 245747-71-1
CAS RN 245747-71-1
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
NFAT Transcription Factor Regulator-1, MedChemExpress
MedChemExpress NFAT Transcription Factor Regulator-1 is an IL-2 synthesis inhibitor with an IC50 of 182 nM.