CAS RN 24916-51-6
CAS RN 24916-51-6
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Acetylspiramycin, MedChemExpress
MedChemExpress Acetylspiramycin (Spiramycin B; Spiramycin II; Foromacidin B) is a potent and orally active macrolide antibiotic produced by various Streptomyces species, an acetylated derivative of Spiramycin (HY-100593).